Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD195 Mouse anti-Human, Brilliant Violet 711, Clone: 2D7/CCR5, BD

Catalog No. BDB563395 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563395 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563395 Supplier BD Biosciences Supplier No. 563395
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2D7/CCR5 monoclonal antibody specifically binds to the human chemokine receptor, CCR5. CCR5 is also known as CD195. CCR5 is a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the beta chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+CD45RO+CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: Regulated upon Activation Normal T-cell Expressed and Secreted (RANTES), Macrophage Inflammatory Protein-1 (MIP-1) and Monocyte Chemoattractant Protein 2 (MCP-2). Additionally, CCR5 has been found to be a coreceptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD195
Applications Flow Cytometry
Classification Monoclonal
Clone 2D7/CCR5
Conjugate Brilliant Violet 711
Description CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CCR5 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.