Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD195 Rat anti-Mouse, Unlabeled, Clone: C34-3448, BD

Catalog No. BDB559921 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559921 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559921 Supplier BD Biosciences Supplier No. 559921
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The C34-3448 antibody reacts with mouse chemokine receptor CD195 (also known as CCR5), a member of the β - chemokine receptor family. CD195 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1) α and β . In the mouse system, the gene for CD195 has been mapped to chromosome 9. CD195 mRNA is expressed in heart, spleen and liver tissues, and by macrophages and T-lymphocytes. The immunogen used to generate the C34- 3448 hybridoma was a KLH-conjugated peptide consisting of amino acids 9-30 of mouse CD195 (CCR5).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD195
Applications Flow Cytometry
Classification Monoclonal
Clone C34-3448
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CCR5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CCR5 aa. 9-30
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2c κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.