Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD196 Rat anti-Mouse, Alexa Fluor 647, Clone: 140706, BD

Catalog No. BDB561753 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561753 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561753 Supplier BD Biosciences Supplier No. 561753
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 140706 monoclonal antibody reacts with mouse CD196 (CCR6), a CC chemokine receptor that is expressed on B lymphocytes and subsets of T and dendritic cells. The known ligand for CCR6 is CCL20/MIP-3α, and together they regulate the migration and recruitment of antigen-presenting and immunocompetent cells during inflammatory and immunological responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD196
Applications Flow Cytometry
Classification Monoclonal
Clone 140706
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description CCR6, BN-1, CKR-L3, DRY6, KY411, GPR-CY4, STRL22
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CCR6-transfected Rat Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.