Learn More
CD196 Rat anti-Mouse, Brilliant Violet 421, Clone: 140706, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564736
Description
The 140706 monoclonal antibody specifically binds to mouse CD196 (CCR6), a CC chemokine receptor that is expressed on B lymphocytes and subsets of T and dendritic cells. The known ligand for CCR6 is CCL20/MIP-3 α , and together they regulate the migration and recruitment of antigen-presenting and immunocompetent cells during inflammatory and immunological responses.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications
CD196 | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse CCR6-transfected Rat Cell Line | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
140706 | |
CCR6, BN-1, CKR-L3, DRY6, KY411, GPR-CY4, STRL22 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.