Learn More
CD196 Mouse anti-Human, PE, Clone: 11A9, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551773
Description
Reacts with chemokine receptor CCR6. CCR6 is a seven transmembrane g-protein coupled receptor expressed on peripheral blood lymphocytes and in lymphoid tissues including lymph nodes, spleen, thymus, and appendix. It is a receptor for the CC chemokine MIP-3a/LARC. Clone 11A9 cross-reacts with peripheral blood lymphocytes of rhesus and cynomolgus macaque monkeys. The staining pattern observed shows less reactivity on CD20+ cells than seen on normal human peripheral blood lymphocytes. CCR6 had been clustered as CD196 in the VIIIth HLDA workshop.

Specifications
CD196 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD196/CCR6 Peptide | |
50 Tests | |
Immunology | |
Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
11A9 | |
BN-1; C-C chemokine receptor type 6; C-C CKR-6; CC-CKR-6; CCR-6; CD196 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.