Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD197 (CCR7) Rat anti-Human, Alexa Fluor 647, Clone: 3D12, BD

Catalog No. BDB560921 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560921 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560921 Supplier BD Biosciences Supplier No. 560921
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody 3D12 reacts with the human CC chemokine receptor, CCR7. CCR7 (previously known as BLR-2, EBI-1 and CMKBR7), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MIP-3β/Exodus 3/ELC/ CCL19 and 6Ckine/Exodus 2/SLC/TCA4/CCL21. It has been shown that CCR7 mRNA is expressed mainly in lymphoid tissues including spleen, lymph nodes and tonsil. CCR7 mRNA was also detected in peripheral T and B lymphocytes, in bone marrow and cord blood CD34-positive cells and mature dendritic cells. The human CCR7 gene, unlike other CC chemokine receptor genes, has been mapped to chromosome 17q12. The immunogen used to generate 3D12 hybridoma was the N-terminus as well as parts of the second extracellular loop of human CCR7 protein. The monoclonal antibody 3D12 recognizes an epitope mapping to the N-terminus of human CCR7.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD197
Applications Flow Cytometry
Classification Monoclonal
Clone 3D12
Conjugate Alexa Fluor 647
Description CCR7, BLR-2, EBI-1, CMKBR7
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Human CCR7 protein
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.