Learn More
CD197 (CCR7) Mouse anti-Human, FITC, Clone: 150503, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561271
Description
The monoclonal antibody 150503 specifically binds to the human CC chemokine receptor, CCR7, also known as CD197. CCR7 (previously known as BLR2, EBI1 and CMKBR7), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MIP-3β/Exodus 3/ELC/ CCL19 and 6Ckine/Exodus 2/SLC/TCA4/CCL21. CCR7 mRNA is expressed mainly in lymphoid tissues including spleen, lymph nodes and tonsil. CCR7 is expressed on peripheral T and B lymphocytes, by bone marrow and cord blood CD34-positive cells and by mature dendritic cells. Differential CCR7 expression can be used to distinguish naive, central memory and effector memory T cell subsets. The human CCR7 gene, unlike other CC chemokine receptor genes, has been mapped to chromosome 17 (region 17q12).
Under some complex multi-color conditions, this clone may non-specifically interact with antibodies conjugated with APC-H7 or APC-Cy7, contributing to staining artifacts. BD Horizon Brilliant™ Stain Buffer (Cat. No. 563794), designed to minimize on non-specific fluorescent dye interactions, does not resolve this interaction with either APC-H7 or APC-Cy7. For optimal multicolor staining results, alternatives to APC-H7 and APC-Cy7 should be evaluated.

Specifications
CD197 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human CCR7 Transfected Cell Line | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
150503 | |
CC chemokine receptor 7; BLR2; CMKBR7; EBI1; EVI1; MIP-3 beta receptor | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.