Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD197 (CCR7) Mouse anti-Human, V450, Clone: 150503, BD

Catalog No. BDB560864 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
120 Tests
30 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560864 30 Tests
BDB560863 120 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560864 Supplier BD Biosciences Supplier No. 560864
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 150503 specifically binds to the human CC chemokine receptor, CCR7, also known as CD197. CCR7 (previously known as BLR2, EBI1 and CMKBR7), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MIP-3β/Exodus 3/ELC/ CCL19 and 6Ckine/Exodus 2/SLC/TCA4/CCL21. CCR7 mRNA is expressed mainly in lymphoid tissues including spleen, lymph nodes and tonsil. CCR7 is expressed on peripheral T and B lymphocytes, by bone marrow and cord blood CD34-positive cells and by mature dendritic cells. Differential CCR7 expression can be used to distinguish naive, central memory and effector memory T cell subsets. The human CCR7 gene, unlike other CC chemokine receptor genes, has been mapped to chromosome 17 (region 17q12).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD197
Applications Flow Cytometry
Classification Monoclonal
Clone 150503
Conjugate Violet Fluor 450 (V450)
Description CC chemokine receptor 7; BLR2; CMKBR7; EBI1; EVI1; MIP-3 beta receptor
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CCR7 Transfected Cell Line
Purification Method Affinity Purified
Quantity 30 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.