Learn More
CD1a Mouse anti-Human, PE, Clone: SK9, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 341651
Description
CD1a, clone SK9, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from CD-1 mice immunized with human thymocytes.
CD1a recognizes a human thymocyte antigen, Mr 49 kilodaltons (kd), associated with 12-kd β2 microglobulin.
CD1a recognizes a human thymocyte antigen, Mr 49 kilodaltons (kd), associated with 12-kd β2 microglobulin.

Specifications
CD1a | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human thymocytes | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. | |
IgG2b κ |
Flow Cytometry | |
SK9 | |
PE | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
Purified |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.