Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1b Mouse anti-Human, FITC, Clone: M-T101, BD

Catalog No. BDB555969 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555969 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555969 Supplier BD Biosciences Supplier No. 555969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD1b, a 45 kD membrane glycoprotein expressed in association with β2-microglobulin. CD1b is expressed on cortical thymocytes (moderately), Langerhans cells and interdigitating cells. CD1b has also been reported to react with pyramidal cells in brain. CD1b may participate in a “non-classical” antigen presentation pathway.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1b
Applications Flow Cytometry
Classification Monoclonal
Clone M-T101
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.