Learn More
CD2 Mouse anti-Human, PE, Clone: S5.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347597
Description
CD2, clone S5.2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with T lymphocytes activated by mixed lymphocyte culture.
CD2 recognizes a human lymphocyte antigen, 45 to 50 kilodaltons (kDa), which also forms the binding site for sheep erythrocytes.

Specifications
CD2 | |
Monoclonal | |
6.25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
T lymphocytes activated by mix lymphocyte culture | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
S5.2 | |
PE | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.