Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Rat anti-Mouse, PE, Clone: RM2-5, BD

Catalog No. BDB553112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553112 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553112 Supplier BD Biosciences Supplier No. 553112

Rat Monoclonal Antibody

The RM2-5 monoclonal antibody specifically binds to the immunoglobulin superfamily adhesion molecule, CD2 (LFA-2). CD2 is a type I transmembrane glycoprotein that serves as the the major receptor for CD48 in the mouse. CD2 is involved in T-cell activation, immunoregulation, and thymocyte maturation. In the mouse, CD2 is expressed on peripheral T lymphocytes, B lymphocytes, and NK cells, and a subpopulation of intraepithelial T lymphocytes. CD2 is expressed throughout mouse thymic ontogeny, except for distinct subsets of the CD4-CD8- early thymocytes. In the mouse bone marrow, CD2 is expressed on B220+ sIg+ CD43- pre-B cells, but not on CD43+ pro-B cells. The RM2-5 antibody is one of a set of five anti-mouse CD2 monoclonal antibodies that were classified into two groups according to their mutual competition in binding to cell surface CD2 and which block CD2-mediated cell-cell adhesion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone RM2-5
Concentration 0.2mg/mL
Conjugate PE
Description LFA-2; LFA-3 receptor; Ly37; Ly-37; Lymphocyte antigen 37; T11
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse BALB/c Thymocytes
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b λ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.