Learn More
Description
Host Species: Mouse
Clone: 2H7
Isotype: IgG2b, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human 6.16c1.3 B cell line
Flow cytometry
Specifications
Specifications
| Antigen | CD20 |
| Classification | Monoclonal |
| Clone | 2H7 |
| Conjugate | Brilliant Ultraviolet 395 |
| Host Species | Mouse |
| Immunogen | Human 6.16c1.3 B cell line |
| Purification Method | Affinity Purified |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.