Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20, Mouse anti-Human, Clone: 2H7, BUV395, BD

Catalog No. BDB563782 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563782 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563782 Supplier BD Biosciences Supplier No. 563782
Add to Cart
Add to Cart

Host Species: Mouse
Clone: 2H7
Isotype: IgG2b, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human 6.16c1.3 B cell line

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD20
Classification Monoclonal
Clone 2H7
Conjugate Brilliant Ultraviolet 395
Host Species Mouse
Immunogen Human 6.16c1.3 B cell line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.