Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Mouse anti-Human, PE, Clone: L27, BD

Catalog No. BDB347201 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347201 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347201 Supplier BD Biosciences Supplier No. 347201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD20, clone L27, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with the LB lymphoblastoid cell line.

The CD20 antigen is a phosphoprotein with a molecular weight of 35 or 37 kilodaltons (kd), depending on the degree of phosphorylation. The antigen is not glycosylated.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD20
Applications Flow Cytometry
Classification Monoclonal
Clone L27
Concentration 12.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with BSA and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Lymphoblastoid Cell Line
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.