Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD200, Alexa Fluor 700, Clone: 325516, R&D Systems™
Supplier: R&D Systems FAB27241N
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
CD200/OX2 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
antigen identified by monoclonal MRC OX-2, CD200, CD200 antigenMOX1, CD200 molecule, MOX1, MOX2, MOX2MRC, MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4345 | |
Human | |
Purified |
Flow Cytometry | |
325516 | |
Flow Cytometry 5 uL/10^6 cells | |
P41217.3 | |
CD200 | |
Mouse myeloma cell line NS0-derived recombinant human CD200, Gln31-Gly232, Accession # P41217.3 | |
100 Tests | |
Primary | |
Detects human CD200 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD200 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction