Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200 Mouse anti-Human, Brilliant Violet 605, Clone: MRC OX-104, BD

Catalog No. BDB562853 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562853 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562853 Supplier BD Biosciences Supplier No. 562853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MRC OX-104 monoclonal antibody specifically binds to CD200. CD200 is a 40-45 kDa type 1 transmembrane glycoprotein and is also referred to as OX2. CD200 is a member of the immunoglobulin superfamily of proteins. It contains two Ig domains, a single transmembrane region and a short cytoplasmic tail. CD200 is expressed on resting and PMA-activated T- and B-cells, but not on NK cells, monocytes, granulocytes, or platelets. It has been found on a subset of CD34+ progenitor cells. Interaction of CD200 with its receptor on macrophages induces a downregulation of macrophage activity in a variety of tissues, suggesting a regulatory function.

This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200
Applications Flow Cytometry
Classification Monoclonal
Clone MRC OX-104
Conjugate Brilliant Violet 605
Description OX-2; MOX1; MOX2 ; My033
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD200 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.