Learn More
Description
Host Species: Mouse
Clone MRC OX-104
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Human CD200 Recombinant Protein
Flow Cytometry
Specifications
Specifications
| Antigen | CD200 |
| Classification | Monoclonal |
| Clone | MRC OX-104 |
| Conjugate | Brilliant Violet 510 |
| Host Species | Mouse |
| Immunogen | Human CD200 Recombinant Protein |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.