Learn More
CD200, APC, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA546878
Description
This gene encodes a receptor for the OX-2 membrane glycoprotein. Both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. This receptor is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. Mouse studies of a related gene suggest that this interaction may control myeloid function in a tissue-specific manner. Alternative splicing of this gene results in multiple transcript variants.
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human CD200 is observed.
Specifications
CD200 | |
Polyclonal | |
Buffered saline solution with BSA and <0.1% sodium azide | |
O54901 | |
CD200 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD200 Gln31-Gly232 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
APC | |
Cd200 | |
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
17470 | |
Store at 4°C, store in the dark, do not freeze | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.