Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200 Mouse anti-Human, PerCP-Cy5.5, Clone: MRC OX-104, BD

Catalog No. BDB562124 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562124 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562124 Supplier BD Biosciences Supplier No. 562124
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MRC OX-104 monoclonal antibody specifically binds to CD200. CD200 is a 40-45 kDa type 1 transmembrane glycoprotein and is also referred to as OX2. CD200 is a member of the immunoglobulin superfamily of proteins. It contains two Ig domains, a single transmembrane region and a short cytoplasmic tail. CD200 is expressed on resting and PMA-activated T- and B-cells, but not on NK cells, monocytes, granulocytes, or platelets. It has been found on a subset of CD34+ progenitor cells. Interaction of CD200 with its receptor on macrophages induces a downregulation of macrophage activity in a variety of tissues, suggesting a regulatory function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200
Applications Flow Cytometry
Classification Monoclonal
Clone MRC OX-104
Conjugate PerCP-Cyanine5.5
Description OX-2; MOX1; MOX2 ; My033
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD200 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.