Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD201 Rat anti-Human, Unlabeled, Clone: RCR-252, BD

Catalog No. BDB552500 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552500 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552500 Supplier BD Biosciences Supplier No. 552500

Rat Monoclonal Antibody

Reacts with endothelial cell protein C receptor (EPCR), a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. EPCR is also referred to as CD201. It binds protein C and augments its activation. Reports indicate that EPCR is expressed on several malignant cell lines. Malignant cell lines that express EPCR also show significant levels of protein C activation and this activation could be inhibited by anti-EPCR antibody.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD201
Applications Flow Cytometry
Classification Monoclonal
Clone RCR-252
Concentration 0.5mg/mL
Conjugate Unconjugated
Description EPCR
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.