Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD205 Mouse anti-Human, Alexa Fluor 647, Clone: MMRI-7, BD

Catalog No. BDB558156 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558156 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558156 Supplier BD Biosciences Supplier No. 558156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with DEC-205, a 205 kDa transmembrane protein with C-type lectin external domains, present on dendritic cells (DC) and some epithelia. It is being described as a multilectin receptor for adsorptive endocytosis. Reports indicate that CD205 (DEC-205) mRNA levels increase significantly during activation. In monocyte derived dendritic cells, the expression of CD205 increased during maturation. Antibody MMRI-7 is useful for the study of DC development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD205
Applications Flow Cytometry
Classification Monoclonal
Clone MMRI-7
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description DEC-205
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.