Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD208 Mouse anti-Human, PE, Clone: I10-1112, BD

Catalog No. BDB558126 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558126 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558126 Supplier BD Biosciences Supplier No. 558126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD208, also known as DC-LAMP or lysosome associate membrane protein. DC-LAMP is an intracellular molecule, of approximately 70 - 90 kDa, induced during dendritic cell (DC) maturation. It is undetected in immature DCs, but it is sharply expressed upon activation with LPS, TNFa, or CD154 in DCs from skin, tonsil, cultures of CD34+ cells in presence of GM-CSF and TNFa, and on monocyte cultures in the presence of GM-CSF and IL-4. Reports describe the transient expression of CD208 in the MHC class II compartment before the translocation of the MHC II molecules to the cell surface. It has been suggested that the biological role of DCLAMP could be its participation in peptide loading onto MHC class II.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD208
Applications Flow Cytometry
Classification Monoclonal
Clone I10-1112
Conjugate PE
Description DC-LAMP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.