Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD209a Rat anti-Mouse, APC, Clone: 5H10, BD

Catalog No. BDB564928 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564928 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564928 Supplier BD Biosciences Supplier No. 564928
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 5H10 monoclonal antibody specifically binds to mouse CD209a, or CIRE, which is the most closely related homolog of human DC-SIGN. In the mouse, several homologs of human CD209 (DC-SIGN) have been described. CD209a is a 238-amino acid type-II transmembrane C-type lectin, with an extracellular carbohydrate recognition domain. It can bind to mannosylated ligands in a Ca++ dependent manner. CD209a is expressed by some conventional CD4+ and CD4-CD8- dendritic cells (DC) but not by CD8+ DC. It is also expressed on monocytes, splenic plasmacytoid pre-DC, macrophages, and resident peritoneal cells. CD209a expression is downregulated upon cellular activation. CD209a is thought to play a role in the internalization and presentation of antigens to initiate immune responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD209a
Applications Flow Cytometry
Classification Monoclonal
Clone 5H10
Concentration 0.2mg/mL
Conjugate APC
Description CD209; Cire; DC-SIGN; Dcsign; DC-SIGN1; CDSIGN
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Rat
Immunogen Peptide Derived from Extracellular Region of CD209a/CIRE
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.