Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Mouse anti-Human, Unlabeled, Clone: 1048, BD

Catalog No. BDB552727 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552727 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552727 Supplier BD Biosciences Supplier No. 552727
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD21 is a 145 kDa transmembrane glycoprotein that is expressed on mature B cells, follicular dendritic cells, a subset of immature thymocytes and on some epithelial cells. CD21 is also known as the C3d Receptor, Complement Receptor 2 (CR2), and the Epstein-Barr Virus receptor, (EBV-R). CD21 is part of a large signal-transduction complex that also includes CD19, CD81 and Leu13. CR2 serves as a receptor for the activation fragments of complement component C3, i.e. C3d, iC3d, and C3dg and C3d-bound antigens and plays a central role in the immune response by amplifying B-cell activation and proliferation. CR2 is also the receptor for EBV by binding its surface glycoprotein gp350/220. Anti-human CR2 hybridomas were generated from Cr2-/-mice (lacking the cr2 gene, that encodes CR2 and CR1 in mice) immunized with the recombinant protein representing the first two amino-terminal short consensus repeats (SRC) of human CR2, SCR1-2. Both the C3d and EBV binding sites of hu CR2 were localized to these sites. Clone 1048 has been reported to specifically react with CR2-expressing cells and to inhibit CR2 interactions with C3 fragments or the EBV surface protein, gp350/220.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD21
Applications Flow Cytometry
Classification Monoclonal
Clone 1048
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CR2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.