Learn More
CD21 Mouse anti-Human, Unlabeled, Clone: B-LY4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555421
Description
The B-ly4 monoclonal antibody specifically binds to CD21, a 145 kDa glycosylated type I integral membrane protein. CD21 is a receptor for the C3d complement fragment and for Epstein-Barr virus (EBV). CD21 is expressed on mature B cells, follicular dendritic cells, and some epithelial cells. It is also weakly expressed on the subset of mature T cells and thymocytes. CD21 plays a role in B-cell activation and proliferation. It may also play a role in modulating the function of T cells in the immune response to infections by lymphotropic viruses. Recently, CD21 was found to be part of a large complex containing CD19, CD81, and possibly other molecules.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD21 | |
Monoclonal | |
0.5mg/mL | |
CR2; Complement receptor type 2; C3DR; EBV-R; Epstein-Barr virus receptor | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
B-LY4 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.