Learn More
CD210 Rat anti-Mouse, Unlabeled, Clone: 1B1.3a, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559912
Description
The 1B1.3a antibody reacts with the extracellular region of mouse CD210 which is also known as the mouse IL-10 receptor (mIL-10R); it does not recognize the human IL-10R. The IL-10R is expressed by a variety of mouse cell types and cell lines including thymocytes, T cells, B cells, and monocytes. 1B1.3a is a neutralizing antibody and reportedly blocks the binding of human IL-10, which cross-reacts with the mouse IL-10R. The immunogen used for the generation of the 1B1.3a hybridoma was purified recombinant ligand-binding domain of mIL-10R.

Specifications
CD210 | |
Monoclonal | |
0.5mg/mL | |
IL-10 Receptor | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
1B1.3a | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Purified recombinant ligand-binding domain of mIL-10R | |
0.5 mg | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.