Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD210 Rat anti-Mouse, PE, Clone: 1B1.3a, BD

Catalog No. BDB559914 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559914 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559914 Supplier BD Biosciences Supplier No. 559914
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 1B1.3a antibody reacts with the extracellular region of mouse CD210 which is also known as the mouse IL-10 receptor (mIL-10R); it does not recognize the human IL-10R. The IL-10R is expressed by a variety of mouse cell types and cell lines including thymocytes, T cells, B cells, and monocytes. 1B1.3a is a neutralizing antibody and reportedly blocks the binding of human IL-10, which cross-reacts with the mouse IL-10R. The immunogen used for the generation of the 1B1.3a hybridoma was purified recombinant ligand-binding domain of mIL-10R.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD210
Applications Flow Cytometry
Classification Monoclonal
Clone 1B1.3a
Concentration 0.2mg/mL
Conjugate PE
Description IL-10 Receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified recombinant ligand-binding domain of mIL-10R
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.