Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD212 Mouse anti-Mouse, Biotin, Clone: 114, BD

Catalog No. BDB551973 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551973 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551973 Supplier BD Biosciences Supplier No. 551973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 114 antibody reacts with mouse CD212 (the β1 subunit of IL-12Rβ1), originally termed IL-12Rβ, of the mouse IL-12 receptor complex. The IL-12Rβ1 subunit associates with a β2 subunit to form a heterodimeric IL-12 receptor complex. Each one of the IL-12R subunits exhibits low affinity for IL-12, but in combination, they bind IL-12 with high affinity. The IL-12Rβ1 subunit interacts primarily with IL-12 p40 whereas the IL-12Rββ1 has more recently been described to bind IL-23, a heterodimer formed of the p40 subunit from IL-12, and p19. The cytoplasmic regions of the β1 and β2 subunits contain the box1 and box2 motifs found in other cytokine receptors such as gp130, LIFR and G-CSFR. IL-12Rβ1 are primarily expressed by activated T cells and NK cells. Experiments with IL-12Rβ1 deficient mice have shown that IL-12Rβ1 is necessary for mouse T and NK cell responsiveness to IL-12 p75. The 114 antibody was generated by immunizing IL-12Rβ1 deficient mice of (129 x BALB/c)F1 background with mouse Ba/F3 cells that were stably transfected with IL-12Rβ1

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD212
Applications Flow Cytometry
Classification Monoclonal
Clone 114
Concentration 0.5mg/mL
Conjugate Biotin
Description IL-12 Receptor β1 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IL-12Rβ1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.