Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD212 Rat anti-Human, Unlabeled, Clone: 2B6/12 Tests2, BD

Catalog No. BDB550722 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550722 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550722 Supplier BD Biosciences Supplier No. 550722
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reacts with IL-12R β 2 subunit, one of two subunits of IL-12R β . It is expressed on T cells and NK cells. IL-12R is a glycoprotein member of the hemopoietin receptor superfamily and is related to gp130, the granulocyte-colony stimulating factor (G-CSF) receptor and leukemia inhibitory factor (LIF). Reports indicate that both, IL-12R β 1 (CD212) and IL-12R β 2 subunits together are capable of high affinity binding to IL-12. IL-12 is an immunomodulatory cytokine produced mainly by antigen presenting cells (APC) and plays a role in promoting Th1-type responses and cell mediated immunity. It stimulates proliferation of T cells and NK cells, enhances lytic activity of lymphokine activated (LAK) cells and CTL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD212
Applications Flow Cytometry
Classification Monoclonal
Clone 2B6/12 Tests2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IL-12 receptor subunit beta-2; Interleukin-12 receptor beta-2 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.