Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD212 Mouse anti-Human, PE, Clone: 2.1E6, BD

Catalog No. BDB556065 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556065 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556065 Supplier BD Biosciences Supplier No. 556065
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with IL-12Rβ1 subunit, one of two subunits of the IL-12Rβ. IL-12R, expressed mainly on T cells or NK cells, is a member of the hemopoietin receptor superfamily and is related to gp130, granulocyte-colony stimulating factor (G-CSF) receptor and leukemia inhibitory factor (LIF). Reports indicate that together, subunits IL-12Rβ1 and IL-12Rβ2 bind IL-12 with high affinity. IL-12 is an immunomodulatory cytokine produced mainly by antigen-presenting cells (APC) and plays a role in promoting Th1-type responses and cell-mediated immunity. It stimulates proliferation of T cells and NK cells, enhances lytic activity of lymphokine-activated killer (LAK) cells and CTL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD212
Applications Flow Cytometry
Classification Monoclonal
Clone 2.10E+06
Conjugate PE
Description IL-12R beta 1, IL-12 receptor β1 subunit
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen PHA-activated Human PBMC
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.