Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD218a (IL-18Rα) Mouse anti-Human, PE, Clone: H44, BD

Catalog No. BDB564675 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564675 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564675 Supplier BD Biosciences Supplier No. 564675
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The H44 monoclonal antibody specifically binds to CD218a, which is also known as Interleukin-18 receptor 1 (IL-18R1), IL-18 Receptor alpha (IL-18R α ) and IL1 Receptor-related protein (IL-1Rrp). CD218a is a type I transmembrane glycoprotein that associates with CD218b (IL-18R β ) subunit to form the functional IL-18 Receptor Complex. Both subunits belong to the IL-1R family. The IL-18R α chain is responsible for extracellular IL-18 binding whereas the IL-18R β subunit is involved in intracellular signaling through the NF κ B pathway. CD218a is expressed on a variety of cells including T cells, NK cells, macrophages, granulocytes, keratinocytes, endothelial cells, and smooth muscle cells. CD218a expression is induced weakly in T cells by IL-12 treatment and moderately by phytohemagglutinin (PHA) stimulation. Costimulation with both IL-12 and PHA induces optimal CD218a expression on CD4+ T cells, CD8+ T cells, and CD56+ NK cells. The IL-18/IL-18R Complex plays important roles in adaptive immunity as well as innate immune responses and autoimmunity. H44 reportedly neutralized the biological activity of recombinant human IL-18 protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD218a (IL-18Rα)
Applications Flow Cytometry
Classification Monoclonal
Clone H44
Conjugate PE
Description IL-1Rrp; IL1RRP; IL18RA; IL-18Rα; IL-18R1; IL-18R-1; IL18R1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.