Learn More
CD22 Mouse anti-Human, APC, Clone: S-HCL-1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340933
Description
The CD22 antibody, clone S-HCL-1, is derived from the hybridization of NS-1 mouse myeloma cells with spleen cells isolated from CD-1 mice immunized with whole hairy cell leukemia cells and membrane preparations derived from them.
The CD22 antibody recognizes a 135-kilodalton (kDa) type I transmembrane glycoprotein in the immunoglobulin superfamily. The CD22 antigen is also known as BL-CAM, Bgp135, and Siglec2.

Specifications
CD22 | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Whole hairy cell leukemia cells and membrane preparation | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
S-HCL-1 | |
APC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.