Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD22 (BL-CAM) Mouse anti-Human, FITC, Clone: S-HCL-1, BD

Catalog No. BDB347573 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347573 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347573 Supplier BD Biosciences Supplier No. 347573
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD22 antibody, clone S-HCL-1, is derived from the hybridization of NS-1 mouse myeloma cells with spleen cells isolated from CD-1 mice immunized with whole hairy cell leukemia cells and membrane preparations derived from them.

The CD22 antibody recognizes a 135-kilodalton (kDa) type I transmembrane glycoprotein in the immunoglobulin superfamily. The CD22 antigen is also known as BL-CAM, Bgp135, and Siglec2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD22 (BL-CAM)
Applications Flow Cytometry
Classification Monoclonal
Clone S-HCL-1
Concentration 12.5μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Whole hairy cell leukemia cells and membrane preparation
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.