Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD220 Mouse anti-Human, Unlabeled, Clone: 3B6/IR, BD

Catalog No. BDB559954 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559954 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559954 Supplier BD Biosciences Supplier No. 559954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with insulin receptor (IR), a glycoprotein composed of two extracellular α-subunits (130 kDa) and two transmembrane β-subunits (95 kDa). It is expressed on human hematopoietic and non-hematopoietic cells, but unlike the insulin-like growth factor receptor (IGF-IR), which is ubiquitous, the insulin receptor is restricted to major target tissues of insulin action. Upon binding insulin, the IR subunits form a heterotetramer of two α and two β subunits resulting in autophosphorylation and activation of the tyrosine kinase activity of the receptor. This ligand-receptor interaction is important in regulating cell metabolism and growth. 3B6/IR monoclonal antibody reacts similarly to anti-human IR α, clone 47-9, an α-subunit antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD220
Applications Flow Cytometry
Classification Monoclonal
Clone 3B6/IR
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Insulin Receptor, Insulin R
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.