Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD220 Mouse, Unlabeled, Clone: 46, BD

Catalog No. BDB610109 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610109 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610109 Supplier BD Biosciences Supplier No. 610109
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin Receptor (IR) is a transmembrane receptor tyrosine kinase which, upon insulin binding, initiates a cascade of events, including autophosphorylation, phosphorylation of cellular protein substrates, glucose transport, and glycogen synthesis. IR is synthesized as a large glycosylated precursor that is cleaved upon maturation into a 130kDa α-subunit with kinase activity and a 95kDa β-subunit. The active Insulin Receptor is a heterotetramer of homologus α and β subunits joined by disulfide bonds. Among the major cytosolic substrates of the Insulin Receptor are IRS-1 and -2, β-Adrenergic receptor, and pp15 (adipocyte lipid-binding protein, ALBP). Autophosphorylation of the IR recruits IRS-1 and -2 to the phosphotyrosines. Subsequently, the phosphorylated IRS-1 and -2 act as docking sites for other signaling proteins like PI3-Kinase, Shc, PTP1D, Nck, etc. In addition, the phosphatase LAR is tightly associated with the IR and LAR becomes activated after insulin stimulation dephosphorylating the IR and its substrates. Therefore, LAR provides a turn-off mechanism in insulin signaling.

Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Human Insulin pro-receptor β subunit 1006-1144
Formula Weight [Chemical]: 95kDa

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD220
Applications Western Blot
Classification Monoclonal
Clone 46
Concentration 250μg/mL
Conjugate Unconjugated
Description Insulin Receptor β
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Insulin pro-receptor β subunit 1006-1144
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.