Learn More
Description
Reacts with insulin receptor (IR), a glycoprotein composed of two extracellular α -subunits (130 kDa) and two transmembrane β -subunits (95 kDa). It is expressed on human hematopoietic and non-hematopoietic cells, but unlike the insulin-like growth factor receptor (IGF-IR), which is ubiquitous, the insulin receptor is restricted to major target tissues of insulin action. Upon binding insulin, the IR subunits form a heterotetramer of two α and two β subunits resulting in autophosphorylation and activation of the tyrosine kinase activity of the receptor. This ligand-receptor interaction is important in regulating cell metabolism and growth. 3B6/IR monoclonal antibody reacts similarly to anti-human IR α , clone 47-9, an α -subunit antibody.
Specifications
Specifications
| Antigen | CD220 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3B6/IR |
| Conjugate | PE |
| Description | Insulin Receptor, Insulin R |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.