Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD221 Mouse anti-Human, Unlabeled, Clone: 3B7, BD

Catalog No. BDB556000 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556000 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556000 Supplier BD Biosciences Supplier No. 556000

Mouse Monoclonal Antibody

Reacts with human insulin-like growth factor-I (IGF-I) receptor. IGF-I receptor is a glycoprotein complex composed of two extracellular α subunits and two transmembrane β subunits. The transmembrane subunits play a role in tyrosine phosphorylation of several intracellular signaling proteins. IGF-I receptor is structurally and functionally similar to insulin receptor and is expressed on a variety of human hematopoietic and non-hematopoietic cells. 3B7 antibody does not cross react with human insulin receptor; it stimulates IGF-I and IGF-II induced cell growth and activates autophosphorylation of the IGF-I receptor β subunit.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD221
Applications Flow Cytometry
Classification Monoclonal
Clone 3B7
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IGF-I receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.