Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD221 (pY1131) Mouse, Alexa Fluor 647, Clone: K74-218, BD

Catalog No. BDB558588 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558588 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558588 Supplier BD Biosciences Supplier No. 558588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Insulin-like growth factor-1 (IGF-1) receptor, or CD221, is a receptor tyrosine kinase that has high affinity for IGF-1 and low affinity for insulin and IGF-2. Each transmembrane protein complex is formed from a pair of 1367 amino-acid precursor proteins. After the 30 amino-acid signal sequences are removed, the paired polypeptides are cleaved to derive an a and b chain from each. The two extracellular a chains form the ligand-binding domain, while the transmembrane-and-intracellular b chains carry the kinase activity. Upon stimulation by its ligand, IGF-1 receptor autophosphorylates multiple tyrosine sites in its b chains and phosphorylates other signaling mediators that regulate cell growth, development, and neoplastic transformation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD221 (pY1131)
Applications Flow Cytometry
Classification Monoclonal
Clone K74-218
Conjugate Alexa Fluor 647
Description IGF1R; IGF-I receptor; IGFIR; IGFR; Insulin-like growth factor 1 receptor
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human IGF-1 Receptor Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.