Learn More
CD227 Mouse anti-Human, Unlabeled, Clone: HMPV, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555925
Description
Reacts with the core peptide of the MUC1 protein, a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (>200kDa) and an amino acid composition rich in serine, threonin, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats which function as multiple epitopes for the monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in staining intensity between normal and malignant tissues. This antibody has been shown to react with both normal and malignant epithelia of various tissues including breast and colon.

Specifications
CD227 | |
Monoclonal | |
0.5mg/mL | |
MUC1; CA 15-3; DF3; EMA; Episialin; H23; H23AG; KL-6; MAM6; PEM; PEMT; PUM | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
HMPV | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Milk Fat Globule Membranes | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.