Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD229 Mouse anti-Human, PE, Clone: HLy9.1.25, BD

Catalog No. BDB565238 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565238 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565238 Supplier BD Biosciences Supplier No. 565238
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HLY9.1.25 monoclonal antibody specifically binds to CD229 which is also known as, Signaling lymphocyte activation molecule family member 3 (SLAMF3), Lymphocyte antigen 9 (Ly9), or T-lymphocyte surface antigen Ly-9. CD229 is a 100-120 kDa, single-pass type I membrane glycoprotein that belongs to the SLAM family within the Ig superfamily. CD229 is expressed on thymocytes, T cells, B cells, dendritic cells, and NK cells. Through homophilic binding, CD229 may play a role in adhesive interactions between T and B cells. During T cell activation, CD229 localizes to the immunological synapse.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD229
Applications Flow Cytometry
Classification Monoclonal
Clone HLy9.1.25
Conjugate PE
Description SLAMF3; SLAM family member 3; LY9; Ly-9; hly9
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human hLy9/CD229 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.