Learn More
Description
Host Species: Rat
Clone B3B4
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen FcΕR isolated from the mouse B hybridoma line O1.2B2
Flow Cytometry
Specifications
Specifications
| Antigen | CD23 |
| Classification | Monoclonal |
| Clone | B3B4 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 647 |
| Host Species | Rat |
| Immunogen | FcΕR isolated from the mouse B hybridoma line O1.2B2 |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.