Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23 Mouse anti-Human, Unlabeled, Clone: M-L233, BD

Catalog No. BDB555707 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555707 0.1 mg
BDB555708 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555707 Supplier BD Biosciences Supplier No. 555707

Mouse Monoclonal Antibody

The M-L233 antibody reacts with the low affinity receptor for human IgE, FcεRII/CD23. CD23 is a type II membrane protein which can be expressed by B cells, monocytes, macrophages, eosinophils, platelets, and dendritic cells. CD23 can mediate IgE-dependent cytotoxicity and phagocytosis by macrophages and eosinophils. Soluble CD23 (sCD23) can be released by CD23+ cells as a result of proteolytic cleavage of membrane CD23. Larger fragments of sCD23 (e.g., 37-25kDa) retain their IgE-binding capacity whereas smaller fragments (i.e., <12kDa) do not. Soluble CD23 may have immunoregulatory effects on the growth and differentiation of B cells and other cell types.

Host Species: Mouse
Clone: M-L233
Isotype: IgG1 κ
Species Reactivity: Human

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD23
Applications Flow Cytometry
Classification Monoclonal
Clone M-L233
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.