Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23 Mouse anti-Human, Unlabeled, Clone: M-L233, BD

Catalog No. BDB550386 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550386 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550386 Supplier BD Biosciences Supplier No. 550386

Mouse Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD23
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone M-L233
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.