Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD235a Mouse anti-Human, FITC, Clone: GA-R2 (HIR2), BD

Catalog No. BDB561017 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561017 25 μg
BDB559943 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561017 Supplier BD Biosciences Supplier No. 561017
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The GA-R2 (also known as HIR2) monoclonal antibody specifically binds to CD235a, otherwise known as Glycophorin A (GYPA, GPA), PAS-2, MN sialoglycoprotein (MNS, MN), and Sialoglycoprotein A/alpha. CD235a is a type I transmembrane sialoglycoprotein that is expressed on human erythrocytes, erythroid precursor cells and certain leukemic cell types. This antibody is useful for the identification and characterization of erythrocytes, certain myeloid leukemic cell types, and studies of erythroid cell development and infectious diseases with erythrocyte involvement. Mature, non-nucleated red blood cells characteristically express very high levels of glycophorin A.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD235a
Applications Flow Cytometry
Classification Monoclonal
Clone GA-R2 (HIR2)
Concentration 0.5mg/mL
Conjugate FITC
Description Glycophorin-A; GYPA; GPA; PAS-2; MN sialoglycoprotein; Sialoglycoprotein A
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.