Learn More
CD24 Mouse anti-Rat, Unlabeled, Clone: HIS50, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551133
Description
The HIS50 monoclonal antibody specifically binds to CD24 (Heat-Stable Antigen, HSA or HsAg), a variably glycosylated GPI-anchored membrane protein. In the rat, Cd24 mRNA is found in a variety of epithelia, neural tissue, lymph node germinal centers and thymus. The HIS50 antibody detects CD24 on most B-lineage cells in the bone marrow, on most highly surface IgM (sIgM)-positive B lymphocytes in the blood and spleen, and on marginal-zone B cells in the spleen and lymph node. Variable levels of CD24 are detected on B cells that expresses low levels of sIgM and on follicular B lymphocytes. The HIS50 antibody does not detect CD24 on rat thymocytes nor peripheral T lymphocytes, NK cells, or granulocytes. The HIS50 antibody crossreacts with intracellular antigen(s) in human lymphocytes.
Specifications
| CD24 | |
| Monoclonal | |
| 0.5mg/mL | |
| HSA; Heat-stable antigen; Nectadrin; Cd24a; Signal transducer CD24 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| HIS50 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat bone marrow cells | |
| 0.1 mg | |
| Immunology | |
| Rat | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.