Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD244.1 Rat anti-Mouse, Unlabeled, Clone: C9.1, BD

Catalog No. BDB552421 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552421 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552421 Supplier BD Biosciences Supplier No. 552421

Rat Monoclonal Antibody

CD244 (also known as 2B4 Antigen) is a member of the CD2 subset of the immunoglobulin superfamily (CD2 IgSF).3,4,5 It is expressed on all natural killer (NK) cells, IL-2–activated NK cells (LAK), and a subset of T lymphocytes which mediate non-MHC–restricted cytotoxicity. The C9.1 antibody reacts with CD244.1, the 2B4 alloantigen which is expressed in most strains tested (A/J, AKR/N, BALB/c, CBA/N, C3H/He, DBA/1, DBA/2, NZB/N, SJL/J, 129/Svj), but not C57BL/6 or C58/J mice. The expression of the CD244.1 alloantigen on B lymphocytes that was originally described, has not been confirmed in later studies. The extracellular domain of CD244 is a ligand for another CD2 IgSF member, CD48.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD244.1
Applications Flow Cytometry
Classification Monoclonal
Clone C9.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description 2B4 BALB Alloantigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse MHC-unrestricted killer hybridoma 7D4
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.