Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD244 Mouse anti-Human, PE, Clone: 2-69, BD

Catalog No. BDB550816 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550816 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550816 Supplier BD Biosciences Supplier No. 550816
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2-69 monoclonal antibody specifically binds to 2B4 which is also known as CD244. CD244 is an approximately 63-70 kDa type I transmembrane glycoprotein. It is a SLAM submember (SLAMF4) and CD2 family member of the immunoglobulin superfamily. CD244 was originally identified in the mouse as a non-MHC-restricted cytotoxicity mediator present on NK cells and CD8+ T cells. In humans, CD244 is expressed on NK cells, γδ T cells, subsets of effector and memory CD8+ T cells, monocytes, eosinophils and basophils. This expression pattern suggests a broad role for CD244 in the regulation of leukocyte activation. CD244 binds CD48 with high affinity. CD244 may serve both stimulatory and inhibitory functions depending on a number of factors. These include the intensity and duration of CD244 interactions with its ligands as well as the presence of other signals generated through different receptor-ligand interactions. 2B4 was clustered as CD244 in the VIIth HLDA workshop.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD244
Applications Flow Cytometry
Classification Monoclonal
Clone Feb-69
Conjugate PE
Description 2B4; SLAMF4; NAIL; NK cell activation inducing ligand ; NKR2B4; Nmrk
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.