Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD247 Mouse anti-Mouse, Unlabeled, Clone: 1Z3A1, BD

Catalog No. BDB554241 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554241 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554241 Supplier BD Biosciences Supplier No. 554241

Mouse Monoclonal Antibody

The T-cell receptor (TCR), expressed by thymus-derived lymphocytes, is a multicomponent complex responsible for recognizing antigen in the context of MHC molecules. Ti is the antigen-specific binding component of the TCR. On the majority of peripheral T lymphocytes, Ti is present as a disulfide-linked α - β heterodimer. A minor fraction express Ti as a γ- δ heterodimer. The Ti subunits are Ig-like, each containing V and C regions. Ti is noncovalently associated with an invariant set of molecules referred to as the CD3 subunits. CD3 appears early in thymocyte differentiation and remains expressed on all mature T lymphocytes. In the mouse, it consists of five chains: γ, δ , ε (epsilon), ζ (zeta), and p21. The relative masses of the mouse CD3 chains are 21, 28, 26, 32 (dimer), and 21 kDa respectively.

3A1 recognizes an epitope in the C-terminus of mouse CD3 ζ . A synthetic peptide corresponding to the C-terminus of mouse CD3 ζ (BSACDTYDALHMQTLAPR) was used as immunogen. Hybridomas were derived from Balb/c spleen cells x NS-1.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD247
Applications Western Blot
Classification Monoclonal
Clone 1Z3A1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CD3ζ chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse CD3ζ C-terminus synthetic peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.