Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD247 with control Mouse anti-Human, Unlabeled, Clone: 8D3, BD

Catalog No. BDB551033 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551033 50 μg
BDB551034 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB551033 Supplier BD Biosciences Supplier No. 551033
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The T-cell receptor (TCR), expressed by thymus-derived lymphocytes, is a multi-component complex responsible for recognizing antigen in the context of MHC molecules. Ti is the antigen-specific binding component of the TCR. On the majority of peripheral T lymphocytes Ti is present as a disulfide-linked α - β heterodimer. A minor fraction express Ti as a γ- δ heterodimer. The Ti subunits are Ig-like, each containing V and C regions. Ti is noncovalently associated with an invariant set of molecules referred to as the CD3 subunits. CD3 appears early in thymocyte differentiation and remains expressed on all mature T lymphocytes. In the mouse, it consists of five chains, γ(gamma), δ (delta), ε (epsilon), ζ (zeta), and p21. The relative masses of the mouse CD3 chains are 21, 28, 26, 32 (dimer), and 21 kDa respectively.

The 8D3 antibody recognizes the ζ chain of the human and mouse T cell antigen receptor-associated CD3 complex. It reacts with 16 kDa unphosphorylated monomers and 32 kDa unphosphorylated dimers. It does not appear to recognize 21 kDa phosphorylated monomers or 42 kDa phosphorylated dimers. A synthetic peptide corresponding to a region of human CD3 ζ was used as immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD247 with control
Applications Western Blot
Classification Monoclonal
Clone 8D3
Conjugate Unconjugated
Description CD3ζ chain
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Includes Purified Mouse Anti-Human CD247 (50μg), Jurkat Cell Lysate (50μg)
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.